Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B7-H2/ICOSLG Mouse anti-Human, Janelia Fluor™ 646, Clone: OTI2C5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271735JF646

Catalog No. N271735J646


Add to cart

Description

Description

B7-H2/ICOSLG Monoclonal antibody specifically detects B7-H2/ICOSLG in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

B7-H2/ICOSLG
OTI2C5
Immunohistochemistry
Affinity Purified, Conjugated
ICOSLG
Full length recombinant protein of human B7-H2/ICOSLG (NP_056074) produced in HEK293T cells.
Immunogen affinity purified
Cell Cycle and Replication
Primary
23308.0
Immunohistochemistry
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
B7H2B7 homolog 2, B7-H2B7-related protein 1, B7RP1B7-like protein Gl50, B7RP-1LICOS, CD275, CD275 antigen, GL50transmembrane protein B7-H2 ICOS ligand, ICOSL, ICOS-L, inducible T-cell co-stimulator ligand, KIAA0653ICOS ligand
Mouse
IgG2a
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit