Learn More
B7-H2/ICOSLG Rat anti-Mouse, Alexa Fluor™ 488, Clone: 987602, R&D Systems™
Rat Monoclonal Antibody
Manufacturer:  R&D Systems FAB10083G100UG
Catalog No. FB10083G100
Add to cart
This item is not returnable. View return policy
Description
Description
B7-H2/ICOSLG Monoclonal antibody specifically detects B7-H2/ICOSLG in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
B7-H2/ICOSLG
987602
Flow Cytometry 0.25-1 ug/10^6 cells
Q9JHJ8
B7H2, B7H2B7 homolog 2, B7-H2B7-related protein 1, B7RP-1, B7RP1B7-like protein Gl50, B7RP-1LICOS, CD275, CD275 antigen, GL50, GL50transmembrane protein B7-H2 ICOS ligand, ICOSL, ICOS-L, ICOSLG, inducible T-cell co-stimulator ligand, KIAA0653ICOS ligand
Mouse myeloma cell line NS0-derived recombinant mouse B7-H2, Met1-Lys279, Accession No. Q9JHJ8
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects mouse B7-H2 in direct ELISAs.
Flow Cytometry
Alexa Fluor™ 488
Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide.
Purified, Supernatant, Conjugated
Rat
IgG2a
100 μg
Primary
23308.0
Mouse