Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B7-H2/ICOSLG Antibody (599829) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals MAB1581
Description
B7-H2/ICOSLG Monoclonal specifically detects B7-H2/ICOSLG in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
B7-H2/ICOSLG | |
Monoclonal | |
1.0 mg/mL | |
Flow Cytometry, CyTOF-ready | |
B7H2B7 homolog 2, B7-H2B7-related protein 1, B7RP1B7-like protein Gl50, B7RP-1LICOS, CD275, CD275 antigen, GL50transmembrane protein B7-H2 ICOS ligand, ICOSL, ICOS-L, inducible T-cell co-stimulator ligand, KIAA0653ICOS ligand | |
Rat | |
Protein A or G purified | |
RUO | |
23308 | |
Store at -20C in the dark. Avoid freeze-thaw cycles. | |
IgG1 |
Flow Cytometry, CyTOF | |
599829 | |
Unconjugated | |
PBS with No Preservative | |
ICOSLG | |
rmB7-H2 aa 47-279 | |
100 μg | |
Primary | |
Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction