Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B7-H2/ICOSLG Rat anti-Mouse, Clone: 599829, Novus Biologicals™

Rat Monoclonal Antibody

$175.82 - $436.00

Specifications

Antigen B7-H2/ICOSLG
Clone 599829
Concentration 1.0 mg/mL
Dilution Flow Cytometry, CyTOF-ready
Applications Flow Cytometry, CyTOF
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB1581SP
View Documents
Novus Biologicals
MAB1581SP
25 μg
Each for $175.82
Add to cart
 
MAB1581
View Documents
Novus Biologicals
MAB1581
100 μg
Each for $436.00
Add to cart
 
Description

Description

B7-H2/ICOSLG Monoclonal specifically detects B7-H2/ICOSLG in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications

Specifications

B7-H2/ICOSLG
1.0 mg/mL
Flow Cytometry, CyTOF
Unconjugated
Rat
Mouse
B7H2B7 homolog 2, B7-H2B7-related protein 1, B7RP1B7-like protein Gl50, B7RP-1LICOS, CD275, CD275 antigen, GL50transmembrane protein B7-H2 ICOS ligand, ICOSL, ICOS-L, inducible T-cell co-stimulator ligand, KIAA0653ICOS ligand
ICOSLG
IgG1
Protein A or G purified
599829
Flow Cytometry, CyTOF-ready
Monoclonal
Purified
RUO
PBS with No Preservative
23308
rmB7-H2 aa 47-279
Primary
Store at -20C in the dark. Avoid freeze-thaw cycles.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit