Learn More
B7-H3/CD276 Mouse anti-Human, Alexa Fluor 647, Clone: MM0104-20J12, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP212048AF647
Catalog No. NB212048647
Add to cart
Description
Description
B7-H3/CD276 Monoclonal specifically detects B7-H3/CD276 in Human samples. It is validated for Western Blot, Product Image.
Specifications
Specifications
B7-H3/CD276
MM0104-20J12
Western Blot
Purified
CD276
Human CD276
Protein G purified
Store at 4C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
B7H34Ig-B7-H3, B7-H3B7 homolog 3, CD276 antigen, CD276 molecule, Costimulatory molecule
Mouse
IgG1
0.1 ml
Primary
80381