Learn More
B7-H6 Mouse anti-Cynomolgus Monkey, Alexa Fluor™ 700, Clone: 1002932, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB10127N100UG
Catalog No. FB10127N100
Add to cart
This item is not returnable. View return policy
Description
Description
B7-H6 Monoclonal antibody specifically detects B7-H6 in Cynomolgus Monkey samples. It is validated for Flow Cytometry.
Specifications
Specifications
B7-H6
1002932
Flow Cytometry 0.25-1 μg/106 cells
B7 homolog 6, B7H6, B7-H6, DKFZp686I21167, DKFZp686O24166, NCR3LG1
Human embryonic kidney cell, HEK293-derived cynomolgus Monkey B7-H6, Asp25-Ser262
100 μg
Primary
374383
Cynomolgus Monkey
Flow Cytometry
Alexa Fluor™ 700
Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide
Mouse
IgG2b
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects Cynomolgus Monkey B7-H6 in direct ELISAs. In direct ELISAs, no cross-reactivity with human B7-H6 is observed.