Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B7-H6 Mouse anti-Human, Clone: B7H6/4821, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen B7-H6
Clone B7H6/4821
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122992
View Documents
Novus Biologicals
NBP30775920UG
20 μg
Each for $138.37
Add to cart
 
NB122991
View Documents
Novus Biologicals
NBP30775910UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

B7-H6 Monoclonal antibody specifically detects B7-H6 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

B7-H6
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
Human
B7 homolog 6, B7-H6, DKFZp686O24166
Recombinant fragment (around aa38-169) of human B7-H6 protein (exact sequence is proprietary)
Monoclonal
Protein A or G purified
B7H6/4821
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Mouse
Cancer, Immunology
10 mM PBS with 0.05% BSA
374383
IgG1 Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit