Learn More
B7-H6 Rat anti-Human, Alexa Fluor™ 647, Clone: 1A5, BD Pharmingen™
Rat Monoclonal Antibody
Manufacturer:  BD Biosciences 566733
Catalog No. BDB566733
Add to cart
Description
Description
B7-H6 Monoclonal antibody specifically detects B7-H6 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
B7-H6
1A5
Alexa Fluor 647
Rat
50 μg
Refrigerate (2 to 8°C)
Monoclonal
Human
Flow Cytometry
0.2 mg/mL
Conjugated, Purified
IgM, κ
RUO
Primary
B7-H6