Learn More
B7-H7/HHLA2 Mouse anti-Human, Alexa Fluor 405, Clone: 907812R, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB80841RV100UG
Catalog No. FAB80841RV1
Add to cart
This item is not returnable. View return policy
Description
Description
B7-H7/HHLA2 Monoclonal antibody specifically detects B7-H7/HHLA2 in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
B7-H7/HHLA2
907812R
Flow Cytometry 0.25-1 ug/10^6 cells
Q9UM44
B7H7, HERV-H LTR-associating 2, HERV-H LTR-associating protein 2, HHLA2, Human endogenous retrovirus-H long terminal repeat-associating protein 2
Human embryonic kidney cell line HEK293-derived human B7-H7/HHLA2, Met1-Asn344, Accession # Q9UM44
Protein A or G purified from cell culture supernatant
RUO
Primary
11148
Flow Cytometry
Alexa Fluor 405
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Purified
Mouse
IgG1
100 μg
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Human