Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAALC Rabbit anti-Human,Mouse,Rat, KHL, Polyclonal, MilliporeSigma™

Rabbit Polyclonal Antibody

$147.00 - $372.00

Specifications

Antigen BAALC
Applications Western Blot, Immunohistochemistry (Paraffin)
Conjugate KHL
Format Affinity Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ABN224310MI
View Documents
MilliporeSigma
ABN2243100UG
100 μg
Each for $372.00
Add to cart
 
ABN224325MI
View Documents
MilliporeSigma
ABN224325UG
25 μg
Each for $147.00
Add to cart
 
Description

Description

BAALC Polyclonal antibody specifically detects BAALC in Human,Mouse,Rat samples. It is validated for Western Blotting, Immunohistochemistry (Paraffin).
Specifications

Specifications

BAALC
KHL
Rabbit
Brain and acute leukemia cytoplasmic protein
KLH-conjugated linear peptide corresponding to 16 amino acids from the N-terminal region of human Brain and acute leukemia cytoplasmic protein (BAALC).
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
Western Blot, Immunohistochemistry (Paraffin)
Affinity Purified
Human, Mouse, Rat
BAALC
Polyclonal
Affinity Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit