Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BAAT1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP257563
Description
BAAT1 Polyclonal specifically detects BAAT1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
BAAT1 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence 1-4 ug/ml | |
BAAT1, BRCA1-associated ATM activator 1, BRCA1-associated protein required for ATM activation protein 1, C7orf27, chromosome 7 open reading frame 27, HEAT repeat-containing protein C7orf27, MGC22916 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
BRAT1 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:LSCLGPTHMGPLALGILKLEHCPQALRTQAFQVLLQPLACVLKATVQAPGPPGLLDGTADDATTVDTLLASKSSCAGLLCRTLAHL | |
100 μL | |
Breast Cancer, Cancer, DNA Repair | |
221927 | |
Human | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction