Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bacdown™ Gel No-Rinse Skin Cleansers
GREENER_CHOICESCI_ED

Use anywhere soap and water are not available.

Supplier:  Decon Laboratories, Inc. 7106/EMD

 View more versions of this product

Catalog No. S66873



Explore available promotions
Add to Cart
This item is not returnable. View return policy

Description

Description

  • Nondrying skin cleanser
  • Special formula of 62% ethyl alcohol and gentle emollients to soothe and moisten skin
  • Reduces bacteria count on hands prior to gloving or entering clean environments

Compliance

Meets OSHA requirements for hand washing.

Specifications

Specifications

Hand Cleanser Gel
173°F
Ethyl Alcohol 62% plus emollients
Bottle, one pump
1 Ea.
473.17 mL
Gel
OSHA
Always ready to instantly cleanse hands when soap and Water are not available
<10
16 oz.
Product Suggestions

Product Suggestions

Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Accessories
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.