Learn More
BACH1 Antibody (OTI4E11), DyLight 488, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP270240G
Catalog No. NBP270240G
Add to Cart
Description
Description
BACH1 Monoclonal specifically detects BACH1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
Specifications
BACH1
Monoclonal
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
BACH1
Full length recombinant protein of human BACH1 (NP_996749) produced in HEK293T cells.
0.1 mL
Primary
Human
IgG1
Western Blot, Flow Cytometry, Immunohistochemistry
OTI4E11
Western Blot, Flow Cytometry, Immunohistochemistry
basic region leucine zipper transcriptional regulator BACH1, BTB and CNC homolog 1, BTB and CNC homology 1, basic leucine zipper transcription factor 1, HA2303, transcription regulator protein BACH1, transcription regulator protein10BACH-1
Mouse
Affinity Purified
Breast Cancer, Cancer, DNA Repair
571.0
Store at 4°C in the dark.
Recommendations
Recommendations