Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BACH1 Mouse anti-Human, PE/Cy5.5, Clone: OTI4E11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270240PECY55

Catalog No. N270240PEC5


Add to cart

Description

Description

BACH1 Monoclonal antibody specifically detects BACH1 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

BACH1
OTI4E11
Flow Cytometry
Affinity Purified, Conjugated
BACH1
Full length recombinant protein of human BACH1 (NP_996749) produced in HEK293T cells.
Immunogen affinity purified
Breast Cancer, Cancer, DNA Repair
Primary
571.0
Flow Cytometry
PE-Cyanine5.5
PBS with 0.05% Sodium Azide
basic region leucine zipper transcriptional regulator BACH1, BTB and CNC homolog 1, BTB and CNC homology 1, basic leucine zipper transcription factor 1, HA2303, transcription regulator protein BACH1, transcription regulator protein10BACH-1
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit