Learn More
ZORO TOOLS BACHARACH GAS ANALYZER KIT
Supplier:  ZORO TOOLS G2573304
Encompass
Item: CO2 Analyzer Kit (Carbon Dioxide)Display: AnalogIncludes: Fyrite Analyzer, Aspirator Sampling Assembly and Carrying CaseCarbon Dioxide Concentration: 0 to 20%
Catalog No. NC1715769
Item Discontinued This item has been discontinued by Fisher Scientific. Please to view product availability in your area.