Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bacitracin, Micronized, USP, Spectrum™

Click to view available options
Quantity:
5 MU
Packaging:
Amber Glass Bottle

BA100, 1405-87-4

Spectrum™ Bacitracin, Micronized, USP is a topical antibiotic used to prevent minor skin infections caused by small cuts, scrapes, or burns. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS 1405-87-4
CAS Max % 1
Molecular Formula C66H103N17O16SZn
Packaging Amber Glass Bottle
InChI Key QSNOBVJFKSQBBD-UHFFFAOYNA-N
SMILES [Zn].CCC(C)C(N)C1=NC(CS1)C(=O)NC(CC(C)C)C(=O)NC(CCC(O)=O)C(=O)NC(C(C)CC)C(=O)NC1CCCCNC(=O)C(CC(N)=O)NC(=O)C(CC(O)=O)NC(=O)C(CC2C=NC=N2)NC(=O)C(CC2=CC=CC=C2)NC(=O)C(NC(=O)C(CCCN)NC1=O)C(C)CC
IUPAC Name 4-(2-{[2-(1-amino-2-methylbutyl)-4,5-dihydro-1,3-thiazol-4-yl]formamido}-4-methylpentanamido)-4-[(1-{[18-(3-aminopropyl)-12-benzyl-15-(butan-2-yl)-3-(carbamoylmethyl)-6-(carboxymethyl)-9-[(4H-imidazol-4-yl)methyl]-2,5,8,11,14,17,20-heptaoxo-1,4,7,10,13,16,19-heptaazacyclopentacosan-21-yl]carbamoyl}-2-methylbutyl)carbamoyl]butanoic acid zinc
Molecular Weight (g/mol) 1488.09
Quantity 5 MU