Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bacitracin, Sterile, USP, Spectrum™

Click to view available options
Quantity:
50 KU
Packaging:
Poly Bottle

B1327, 1405-87-4

Spectrum™ Bacitracin, Sterile, USP is an antibiotic for intramuscular administration. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS 1405-87-4
CAS Max % 1
Bacterial Endotoxins 0.01 USP
Ignition Residue 0.005
Molecular Formula C66H103N17O16SZn
Loss on Drying 0.05
Packaging Poly Bottle
InChI Key QSNOBVJFKSQBBD-UHFFFAOYNA-N
SMILES [Zn].CCC(C)C(N)C1=NC(CS1)C(=O)NC(CC(C)C)C(=O)NC(CCC(O)=O)C(=O)NC(C(C)CC)C(=O)NC1CCCCNC(=O)C(CC(N)=O)NC(=O)C(CC(O)=O)NC(=O)C(CC2C=NC=N2)NC(=O)C(CC2=CC=CC=C2)NC(=O)C(NC(=O)C(CCCN)NC1=O)C(C)CC
IUPAC Name 4-(2-{[2-(1-amino-2-methylbutyl)-4,5-dihydro-1,3-thiazol-4-yl]formamido}-4-methylpentanamido)-4-[(1-{[18-(3-aminopropyl)-12-benzyl-15-(butan-2-yl)-3-(carbamoylmethyl)-6-(carboxymethyl)-9-[(4H-imidazol-4-yl)methyl]-2,5,8,11,14,17,20-heptaoxo-1,4,7,10,13,16,19-heptaazacyclopentacosan-21-yl]carbamoyl}-2-methylbutyl)carbamoyl]butanoic acid zinc
Molecular Weight (g/mol) 1488.09
Quantity 50 KU
Show More Show Less