Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Backyard Bugs
A bucket of bugs.
$17.60
Specifications
For Use With (Application) | These colorful creepy, crawly counters turn sorting and counting activities into fun adventures |
---|---|
Format | Hands-on Learning |
Includes | Bugs, grasshopper, bumblebee, beetle, spider, dragonfly and caterpillar, reusable Plastic tub, suggested activities |
Product Type | Backyard Bugs™ Set |
Category | Mathematics |
Alert:
CHOKING HAZARD Small parts. Not for children under 3 yrs.
Includes:
- Bugs: Grasshopper, bumblebee, beetle, spider, dragonfly, and caterpillar
- Reusable plastic tub
- Suggested activities
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
S78997
![]() |
Learning Resources, Inc![]() LER 0457 |
Set of 72 |
Each for $17.60
|
|
Description
Creepy, crawly critters are great for sorting and counting activities. Soft rubber bugs come in six colors and measure approximately 2 in. (5cm).
Specifications
These colorful creepy, crawly counters turn sorting and counting activities into fun adventures | |
Bugs, grasshopper, bumblebee, beetle, spider, dragonfly and caterpillar, reusable Plastic tub, suggested activities | |
Mathematics | |
PreK |
Hands-on Learning | |
Backyard Bugs™ Set | |
12 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title