Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Baclofen, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g
1 kg
Packaging:
Amber Glass Bottle
Poly Bottle
Poly Pail
1134-47-0
C10H12ClNO2
213.66
KPYSYYIEGFHWSV-UHFFFAOYNA-N
4-amino-3-(4-chlorophenyl)butanoic acid
NCC(CC(O)=O)C1=CC=C(Cl)C=C1
1134-47-0
C10H12ClNO2
213.66
KPYSYYIEGFHWSV-UHFFFAOYNA-N
4-amino-3-(4-chlorophenyl)butanoic acid
NCC(CC(O)=O)C1=CC=C(Cl)C=C1

B1049, 1134-47-0, C10H12ClNO2

Spectrum™ Baclofen, USP is a muscle relaxer and an antispastic agent. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Poly Bottle
Quantity 100 g
Ignition Residue 0.003
CAS Max % 1
UN Number UN2811
Percent Purity 98 to 102%