Learn More
Alfa Aesar™ Bactenecin, Bovine
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J66855MB
 View more versions of this product
Catalog No. AAJ66855MB
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
1483.89
Bovine
Lyophilized powder
RUO
Bactenecin, bovine
HPLC
25mg
Arg-Leu-Cys-Arg-Ile-Val-Val-Ile-Arg-Val-Cys-Arg (Disulfide bridge: Cys3-Cys11)
-30° to -10°C