Learn More
BioVision IncSupplier Diversity Partner Bacterial Genomic DNA Isolation Kit: Bacterial Genomic DNA Isolation Kit: Efficient & rapid method to isolate highly pure, intact DNA from bacteria. Useful for PCR, Cloning, DNA Hybridization, and Souther Blotting studies . 100 isolations. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K309100
Bacterial Genomic DNA Isolation Kit: Bacteria are one of the most abundant and diverse organisms on the planet, which take part in numerous critical ecosystem processes. In addition to their medical and ecological importance, bacteria are also used in various industrial applications such as production of enzymes and biofuels. BioVision's bacterial genomic DNA isolation kit provides convenient and simple step-by-step method for isolating quality genomic DNA from gram-negative and gram-positive bacterial species. This kit utilizes enzymatic reactions to release bacterial DNA from the cell. DNA release from the cell is coupled with adsorption of DNA onto a silica spin-column in the presence of high salt concentration, eliminating the use of toxic organic compounds or solvents. DNA purified by this kit is suitable for various downstream molecular biology applications such as PCR, cloning, DNA hybridization, and Southern Blotting BioVision charges an Ice pack fee for each order.
Catalog No. 50-111-9684