Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Affymetrix/USB™ Bacteriological Agar, Ultrapure

Used as a gelling agent in preparation of bacteriological culture media.

Manufacturer:  Affymetrix/USB™ J10906Q1

 View more versions of this product

Catalog No. AAJ10906Q1


Order in Bulk

Request bulk or custom formats

Add to cart

Description

Description

  • Superior transparency
  • Reliable reproducibility
  • Absence of inhibitors capable of hindering optimal development of microorganisms
  • Extraneous matter reduced to minimum
  • Format: White to light cream-colored powder
  • CAS: 9002-18-0
  • MDL Number: MFCD00081288
Specifications

Specifications

White to light cream-colored
Powder
25 lb.
Used as a gelling agent in preparation of bacteriological culture media.
Ultrapure
Plastic Bottle
Bacteriological Agar
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit