Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bacteriological Peptone, Ultrapure, Thermo Scientific™
High grade peptone prepared specifically for general bacteriological purposes.
Supplier: Thermo Scientific J20048P2
Description
- Format: Powder
- CAS: 73049-73-7
- MDL Number: MFCD00131829
Specifications
Powder | |
Bacteriological Peptone | |
Plastic Bottle |
Ultrapure | |
For General Bacteriological Purposes | |
1 lb. |
Safety and Handling
Recommended Storage : Ambient temperatures
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
RUO – Research Use Only
Spot an opportunity for improvement?
Provide Content Correction