Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Bacteriological Peptone, Ultrapure

High grade peptone prepared specifically for general bacteriological purposes.

$56.50 - $850.50

Specifications

Grade Ultrapure
Format Powder
For Use With (Application) For general bacteriological purposes
Packaging Plastic Bottle
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ20048P2 View Documents Thermo Scientific™
J20048P2
1 lb. Each for $56.50
Order in Bulk

Request bulk or custom formats

Add to cart
 
AAJ20048P5 View Documents Thermo Scientific™
J20048P5
5 lb. Each for $194.50
Order in Bulk

Request bulk or custom formats

Add to cart
 
AAJ20048Q1 View Documents Affymetrix/USB™
J20048Q1
25 lb. Each for $850.50
Order in Bulk

Request bulk or custom formats

Add to cart
 
Description

Description

  • Format: Powder
  • CAS: 73049-73-7
  • MDL Number: MFCD00131829
Specifications

Specifications

Ultrapure
Powder
For general bacteriological purposes
Plastic Bottle
Safety and Handling

Safety and Handling

Recommended Storage : Ambient temperatures

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit