Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Bacto™ Proteose Peptone No. 2

Enzymatic digest of protein

Manufacturer:  Gibco™ 212110

 View more versions of this product

Catalog No. 21-211-0


Add to cart

Description

Description

Used in preparing microbiological culture media.
Specifications

Specifications

Microbiological culture media
Storage conditions: 2°C to 25°C.  Store in cool, dry conditions in well-sealed containers.
Powder
Powder
Formulation of culture media for the cultivation of heterotrophic microorganisms
Store in cool, dry conditions in well-sealed containers, 2°C to 25°C
Bacto™
10 kg
Mammalian, Insect, Microbial
Animal origin
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit