Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Bacto™ Proteose Peptone No. 3

Bacto Proteose Peptone No. 3 is used in preparing microbiological and mammalian cell culture media.

$509.50 - $40600.00

Specifications

Certifications/Compliance LUO, MUO
Culture Type Mammalian, Insect, Microbial
Form Powder
With Additives Proteose Peptone No. 3
Product Line Bacto™
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
DF0122-07-6
View Documents
Gibco™
212220
2 kg
Each for $2,140.00
Add to cart
 
DF0122-08-5
View Documents
Gibco™
212230
10 kg
Each for $11,990.00
Add to cart
 
21-169-2
View Documents
Gibco™
211692
50 kg
Each for $40,600.00
Add to cart
 
DF0122-17-4
View Documents
Gibco™
211693
500 g
Each for $509.50
Add to cart
 
Description

Description

  • Bacto Proteose Peptone No. 3 is a member of the Bacto Proteose Peptones, which are enzymatic digests of protein
  • Studies show that no single peptone is the most suitable nitrogen source for every microbiological application
  • These findings led to the development of Bacto Proteose Peptone, Bacto Proteose Peptone No. 2, and Bacto Proteose Peptone No. 3. Bacto Proteose Peptone No. 4 is a spray-dried version of Bacto Proteose Peptone
  • Bacto Proteose Peptone No. 3 is used in preparing microbiological and mammalian cell culture media
  • Store in cool, dry conditions in well-sealed containers at 2°C to 25°C
Specifications

Specifications

LUO, MUO
Powder
Bacto™
Store in cool, dry conditions in well-sealed containers, 2°C to 25°C
Mammalian, Insect, Microbial
Proteose Peptone No. 3
Animal Origin
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit