Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAFF/BLyS/TNFSF13B Mouse anti-Human, Rat, Allophycocyanin/Cy7, Clone: H4-C7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP104265APCCY7

Catalog No. N104265APC7


Add to cart

Description

Description

BAFF/BLyS/TNFSF13B Monoclonal antibody specifically detects BAFF/BLyS/TNFSF13B in Human, Rat samples. It is validated for Flow Cytometry.
Specifications

Specifications

BAFF/BLyS/TNFSF13B
H4-C7
Flow Cytometry
Conjugated, Purified
TNFSF13B
Recombinant human BLyS (134-285aa) purified from E. coli
Protein G purified
Apoptosis, Cell Cycle and Replication, Cytokine Research, Immunology
Primary
10673.0
Human, Rat
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
ApoL related ligand TALL-1, B lymphocyte stimulator, BAFFB-cell activating factor, B-cell-activating factor, BLyS, BLYSB-lymphocyte stimulator, CD257, CD257 antigen, Dendritic cell-derived TNF-like molecule, DTL, TALL-1delta BAFF, TALL1Delta4 BAFF, THANK, TNF and ApoL-related leukocyte expressed ligand 1, TNF- and APOL-related leukocyte expressed ligand 1, TNF homolog that activates apoptosis, TNFSF20, tumor necrosis factor (ligand) superfamily, member 13b, tumor necrosis factor (ligand) superfamily, member 20, tumor necrosis factor ligand superfamily member 13B, tumor necrosis factor superfamily, member 13b, tumor necrosis factor-like protein ZTNF4, ZTNF4
Mouse
IgG3 Kappa
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Anti-human BLyS mAb, clone H4-C7, is derived from hybridization of mouse FO myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human BLyS protein.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit