Learn More
AdipoGen anti-BAFF (mouse), mAb (blocking) (Sandy-2) (preservative free) (100 µg)
SDP
Supplier:  AdipoGen AG20B0063PFC100
Encompass_Preferred
Monoclonal Antibody. Recognizes mouse BAFF. Isotype: Mouse IgG1. Clone: Sandy-2. Applications: FUNC (Blocking), IP. Liquid. In PBS containing 10% glycerol. BAFF is a master regulator of peripheral B cell survival and, together with IL-6, promotes Ig class-switching and plasma cell differentiation. BAFF co-stimulates activated T cells. Increased levels of soluble BAFF have been detected in the serum of patients with various autoimmune diseases, such as Sjögren’s syndrome, rheumatoid arthritis, multiple sclerosis and systemic lupus erythematosus (SLE). Furthermore, BAFF is found in inflammatory sites in which there is lymphoid neogenesis. BAFF levels are elevated in patients with multiple myeloma and B cell chronic lymphoid leukemia (B-CCL).
Catalog No. 50-205-2977
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.