Learn More
BAFFR/CD268 Rabbit anti-Human, Polyclonal, Bioss Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  BiossSupplier Diversity Partner BS2472R
Catalog No. 50-198-4756
Add to cart
Description
Description
Tumor necrosis factor receptor superfamily member 13C (TNFRSF13C), also known as BAFFR, is a protein in humans is encoded by the TNFRSF13C gene. The BAFFR gene is mapped to chromosome 22q13. 1-q13. 31. It has got 184 amino acid transmembrane protein which is 56% identical to the mouse protein. B cell-activating factor (BAFF) enhances B-cell survival in vitro and is a regulator of the peripheral B-cell population. BAFF plays a crucial role in B cell development and can function through receptors other than BCMA.
Specifications
Specifications
BAFFR/CD268
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
TNFRSF13C
Liquid
Rabbit
IgG
100 μL
-20°C
Primary
115650
TNFRSF13C
1 μg/mL
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Q96RJ3
2010006P15Rik; B cell-activating factor receptor; BAFF receptor; Baffr; BAFF-R; B-cell maturation defect; B-cell maturation defect 1; B-cell-activating factor receptor; Bcmd; Bcmd1; Bcmd-1; BLyS receptor 3; BR3; BROMIX; CD268; CVID4; Lvis22; prolixin; RGD1560810; TNF receptor superfamily member 13C; TNFRSF13C; Tumor necrosis factor receptor superfamily member 13C; tumor necrosis factor receptor superfamily, member 13C
KLH conjugated synthetic peptide derived from human BAFFR.
Protein A
RUO
Antibody
Polyclonal
Human