Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAFFR/TNFRSF13C Mouse anti-Human, Clone: BAFFR/1558, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen BAFFR/TNFRSF13C
Clone BAFFR/1558
Concentration 0.2 mg/ml
Dilution Flow Cytometry 1-2 ug/million cells, Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Flow Cytometry, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122561
View Documents
Novus Biologicals
NBP30754410UG
100 μg
Each for $352.89
Add to cart
 
NB122562
View Documents
Novus Biologicals
NBP30754420UG
20 μg
Each for $138.37
Add to cart
 
Description

Description

BAFFR/TNFRSF13C Monoclonal antibody specifically detects BAFFR/TNFRSF13C in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin)
Specifications

Specifications

BAFFR/TNFRSF13C
0.2 mg/ml
Flow Cytometry, Immunohistochemistry (Paraffin)
Purified
RUO
Human
115650
IgG1 Kappa
Primary
Store at 4°C.
BAFFR/1558
Flow Cytometry 1-2 ug/million cells, Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Mouse
Biologically Active Proteins, Cytokine Research, Immunology, Lipid and Metabolism
10 mM PBS with 0.05% BSA
Recombinant full-length human BAFFR/TNFRSF13C protein
Monoclonal
Protein A purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit