Learn More
BAFFR/TNFRSF13C Rat anti-Mouse, Alexa Fluor™ 532, Clone: 7J21, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP222005AF532
Catalog No. NP22205A532
Add to cart
Description
Description
BAFFR/TNFRSF13C Monoclonal antibody specifically detects BAFFR/TNFRSF13C in Mouse samples. It is validated for Flow Cytometry,ELISA,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
BAFFR/TNFRSF13C
7J21
Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
TNFRSF13C
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein TNFRSF13C. Rat monoclonal anti mouse BAFF Receptor Q9D8D0.
0.1 mL
Primary
115650
ELISA, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
Rat
IgG2
Store at 4°C in the dark.
Monoclonal
Mouse