Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BAFFR/TNFRSF13C Rat anti-Mouse, Clone: 7J21, Novus Biologicals™
Rat Monoclonal Antibody has been used in 1 publication
$177.16 - $493.00
Specifications
Antigen | BAFFR/TNFRSF13C |
---|---|
Clone | 7J21 |
Concentration | LYOPH |
Applications | Flow Cytometry, ELISA, Immunohistochemistry (Paraffin), CyTOF |
Classification | Monoclonal |
Description
BAFFR/TNFRSF13C Monoclonal specifically detects BAFFR/TNFRSF13C in Mouse samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
BAFFR/TNFRSF13C | |
LYOPH | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
B cell-activating factor receptor, BAFF receptor, BAFF-R, BAFFRMGC138235, B-cell-activating factor receptor, BLyS receptor 3, BR3, CD268, CD268 antigen, CVID4, tumor necrosis factor receptor superfamily member 13C, tumor necrosis factor receptor superfamily, member 13C | |
TNFRSF13C | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
7J21 | |
Flow Cytometry, ELISA, Immunohistochemistry (Paraffin), CyTOF | |
Unconjugated | |
Rat | |
Biologically Active Proteins, Cytokine Research, Immunology, Lipid and Metabolism | |
Q9D8D0 | |
115650 | |
IgG2 | |
Protein G purified |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title