Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Bafilomycin A1

Click to view available options
Quantity:
1mg
5mg
88899-55-2
C35H58O9
622.84
MFCD06795130
XDHNQDDQEHDUTM-XRCNURIOSA-N
133268469
(3Z,5E,7S,8R,9R,11Z,15R,16S)-16-[(2R,3S,4R)-4-[(2S,4S,5R,6S)-2,4-dihydroxy-5-methyl-6-propan-2-yloxan-2-yl]-3-hydroxypentan-2-yl]-8-hydroxy-3,15-dimethoxy-5,7,9,11-tetramethyl-1-oxacyclohexadeca-3,5,11,13-tetraen-2-one
CC1CC(=CC=CC(C(OC(=O)C(=CC(=CC(C1O)C)C)OC)C(C)C(C(C)C2(CC(C(C(O2)C(C)C)C)O)O)O)OC)C
88899-55-2
C35H58O9
622.84
MFCD06795130
XDHNQDDQEHDUTM-XRCNURIOSA-N
133268469
(3Z,5E,7S,8R,9R,11Z,15R,16S)-16-[(2R,3S,4R)-4-[(2S,4S,5R,6S)-2,4-dihydroxy-5-methyl-6-propan-2-yloxan-2-yl]-3-hydroxypentan-2-yl]-8-hydroxy-3,15-dimethoxy-5,7,9,11-tetramethyl-1-oxacyclohexadeca-3,5,11,13-tetraen-2-one
CC1CC(=CC=CC(C(OC(=O)C(=CC(=CC(C1O)C)C)OC)C(C)C(C(C)C2(CC(C(C(O2)C(C)C)C)O)O)O)OC)C

Specific, potent inhibitor of vacuolar ATPases

Solubility
Soluble in methanol, ethanol, acetone and chloroform. Insoluble in water.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
Beilstein 4730700
Formula Weight 622.83
Physical Form Oil or Film
Color Colorless
Quantity 5mg
Chemical Name or Material Bafilomycin A1

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : RN9781000

storageNote1 : -30° to -10°C