Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bafilomycin A1, Tocris Bioscience™
Click to view available options
Quantity:
10 μg
Description
Highly potent, selective inhibitor of vacuolar H+-ATPases (IC50 = 0.6 - 1.5 nM in bovine chromaffin granules). Selective for v-ATPase over other ATP hydrolyzing enzymes such as F-ATPases and the H+/K+-ATPase (P-ATPase). Thought to inhibit autophagy either by blocking autophagosome-lysosome fusion (in H4IIE cells), or by blocking lysosomal degradation.
Specifications
Type | Bafilomycin A1 |
Formula Weight | 622.84 |
Percent Purity | >98% |
Quantity | 10 μg |
Chemical Name or Material | Bafilomycin A1 |
Safety and Handling
Recommended Storage | Desiccate at -20°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction