Learn More
Andwin Scientific Autoclavable Bags, Nonhazardous Waste, Clear 914×1143 mm (36×45)", 100pack
SDP
Supplier:  Andwin Scientific 14220044
Encompass_Preferred
Made with high-quality polypropylene, the Autoclavable Bags are made to securely hold nonhazardous waste. The bags can resist tears and autoclaving temperatures up to 135C (275F). This product complies with the strength requirements of the 165g dar
Catalog No. NC9346206