Learn More
Nasco Whirl-Pak™ Open Top Sample Bags
Nasco™ Whirl-Pak™ Open Top Sample Bags are strong, unbreakable, and ensure a leakproof closure
Manufacturer:  Nasco B01471WA
Catalog No. 01-812-86
Add to cart
Description
Description
  • Whirl-Pak™ Open Top Sample Bags are strong and leakproof
  • Unlike glass bottles, they are unbreakable which provide easier handling for milk haulers and laboratory technicians
  • Simply whirl the bag three or more times to close, and bend the tabs onto the bag. Or twist the ends together for a leak proof closure
  • Puncture Proof Tabs increase safety by having the tape on the tab extend past the sharp points of the wire
  • Made of polyethylene, they are designed to carry solids or liquids to a laboratory for testing
  • Bags can be frozen to any temperature, including liquid nitrogen temperatures (-346°F/-210°C). Ensure careful handling after freezing
  • Do not exceed temperatures above 180°F/82°C when using Whirl-Pak™ Open Top Sample Bags
Specifications
Specifications
Leakproof, with Puncture Proof Tabs
9 in.
< 180°F
6 in.
532 mL
Polyethylene
Processing and Collection of Sterilized Samples
500/Pk.
< 82°C
18 oz.
23 cm
15 cm