Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BAG5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5118905
Description
Antibody detects endogenous levels of total BAGE5.
It has been hypothesizes that the BAG-5 protein will induce the death of nigral neurons through its predicted interaction with hsp70, which will cause increased protein aggregation and cell death by disinhibition of hsp70's anti-apoptotic function. It is believed that BAG-5 will play an important role in the mechanisms of neuronal death. BAG-5 may also be of interest due to its possible role as a modulator of the hsp70/hsp40 chaperone axis or its possible interaction and coordination of localization/ modulation of other BAG containing proteins via BAG-BAG heterodimerization.
Specifications
BAG5 | |
Polyclonal | |
Unconjugated | |
BAG5 | |
1600025G07Rik; 4930405J06Rik; BAG cochaperone 5; BAG family molecular chaperone regulator 5; Bag5; BAG-5; BAG-family molecular chaperone regulator-5; BAG-family molecular chaperone regulator-5 (BAG-5); BCL2 associated athanogene 5; Bcl-2-associated athanogene 5; BCL2-associated athanogene 5; KIAA0873; LOC522854 protein | |
Rabbit | |
Affinity chromatography | |
RUO | |
366734, 70369, 9529 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q5QJC9, Q8CI32, Q9UL15 | |
BAG5 | |
A synthesized peptide derived from human BAG5(Accession Q9UL15), corresponding to amino acid residues L13-D63. | |
100 μL | |
Primary | |
Human, Mouse, Rat, Monkey | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction