Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAG5 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549696

Catalog No. PA549696


Add to cart

Description

Description

The antibody detects endogenous level of total BAG5 protein.

The protein encoded by this gene is a member of the BAG1-related protein family. BAG1 is an anti-apoptotic protein that functions through interactions with a variety of cell apoptosis and growth related proteins including BCL-2, Raf-protein kinase, steroid hormone receptors, growth factor receptors and members of the heat shock protein 70 kDa family. This protein contains a BAG domain near the C-terminus, which could bind and inhibit the chaperone activity of Hsc70/Hsp70. Three transcript variants encoding two different isoforms have been found for this gene.
Specifications

Specifications

BAG5
Polyclonal
Unconjugated
BAG5
Liquid
BAG5
Fusion protein corresponding to C terminal 350 amino acids of Human Bcl-2-associated athanogene 5
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
ELISA, Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
Q9UL15
KIAA0873
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
9529
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit