Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BAI1 Mouse anti-Human, Alexa Fluor 750, Clone: 480912, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB49691S100UG
This item is not returnable.
View return policy
Description
BAI1 Monoclonal specifically detects BAI1 in Human samples. It is validated for Flow Cytometry.Specifications
BAI1 | |
Monoclonal | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
brain-specific angiogenesis inhibitor 1, FLJ41988, GDAIF | |
HEK293 human embryonic kidney cell line transfected with human BAI1, Ala31-Thr879, Accession # O14514 | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
480912 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
O14514 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
575 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title