Learn More
BAI2 Rabbit anti-Human, Alexa Fluor 594, Clone: 2528B, R&D Systems™
Rabbit Monoclonal Antibody
Manufacturer:  R&D Systems FAB9338T100UG
Catalog No. FAB9338T100
Add to cart
This item is not returnable. View return policy
Description
Description
BAI2 Monoclonal antibody specifically detects BAI2 in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
BAI2
2528B
Flow Cytometry 0.25-1 ug/10^6 cells
O60241-3
brain-specific angiogenesis inhibitor 2, Brain-specific angiongenesis inhibitor-2
Human embryonic kidney cell, HEK293-derived human BAI2, Phe21-Asp911, Accession # O60241-3
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
576
Flow Cytometry
Alexa Fluor 594
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Purified
Rabbit
IgG
100 μg
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Human