Learn More
BAI2 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 196801AP
Catalog No. 50-173-2483
Add to cart
Description
Description
This antibody is specific to BAI2.
BAI1, a p53-target gene, encodes brain-specific angiogenesis inhibitor, a seven-span transmembrane protein and is thought to be a member of the secretin receptor family. Brain-specific angiogenesis proteins BAI2 and BAI3 are similar to BAI1 in structure, have similar tissue specificities and may also play a role in angiogenesis.
Specifications
Specifications
BAI2
0.29 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O60241
BAI2, brain angiogenesis inhibitor 2
Rabbit
IgG
150 μL
-20°C
Primary
576
Immunohistochemistry (Paraffin)
Unconjugated
ADGRB2
Liquid
ADGRB2
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human