Learn More
BAI3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 197891AP
Catalog No. 50-173-2542
Add to cart
Description
Description
This antibody is specific to BAI3.
This p53-target gene encodes a brain-specific angiogenesis inhibitor, a seven-span transmembrane protein, and is thought to be a member of the secretin receptor family. Brain-specific angiogenesis proteins BAI2 and BAI3 are similar to BAI1 in structure, have similar tissue specificities, and may also play a role in angiogenesis.
Specifications
Specifications
BAI3
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O60242, Q80ZF8
BAI3, brain angiogenesis inhibitor 3, KIAA0550
Rabbit
IgG
150 μL
-20°C
Primary
210933, 301309, 577
Western Blot
Unconjugated
ADGRB3
Liquid
ADGRB3
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat