Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAIAP2 Mouse anti-Human, PE-Cyanine7, Clone: OTI 1D9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271603PECY7

Catalog No. N271603PEC7


Add to cart

Description

Description

BAIAP2 Monoclonal antibody specifically detects BAIAP2 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

BAIAP2
OTI 1D9
Flow Cytometry
Affinity Purified, Conjugated
BAIAP2
Full length recombinant protein of human BAIAP2 (NP_006331) produced in HEK293T cells.
Immunogen affinity purified
Signal Transduction
Primary
10458.0
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
BAI1-associated protein 2IRS-58, BAI-associated protein 2, BAP2, brain-specific angiogenesis inhibitor 1-associated protein 2, Fas ligand-associated factor 3, FLAF3, Insulin receptor substrate p53, Insulin receptor substrate p53/p58, Insulin receptor substrate protein of 53 kDa, IRSp53, IRSp53/58, Protein BAP2
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit