Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BAIAP2 Mouse anti-Mouse, Human, Clone: CA-A10-5, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABT1326100UL
Description
BAIAP2 Monoclonal antibody specifically detects BAIAP2 in Mouse, Human samples. It is validated for Dot Blot, Immunocytochemistry, Immunofluorescence, Western Blotting.
Specifications
BAIAP2 | |
Monoclonal | |
Unconjugated | |
Mouse | |
Protein G purified | |
RUO | |
Mouse, Human | |
Purified |
Dot Blot, Immunocytochemistry, Immunofluorescence, Western Blot | |
CA-A10-5 | |
Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
His-tagged full-length human recombinant Brain-specific angiogenesis inhibitor 1-associated protein 2 (BAIAP2) | |
100 μL | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction