Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BAIAP2 Monoclonal Antibody (OTI8G8), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA525648
Description
BAIAP2 Monoclonal antibody specifically detects BAIAP2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
BAIAP2 | |
Monoclonal | |
0.65 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9UQB8 | |
BAIAP2 | |
Full length human recombinant protein of BAIAP2 produced in HEK293 cell | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
OTI8G8 | |
Unconjugated | |
BAIAP2 | |
BAI1 associated protein 2; BAI1-associated protein 2; BAIAP2; BAI-associated protein 2; BAP2; brain-specific angiogenesis inhibitor 1-associated protein 2; Fas ligand-associated factor 3; FLAF3; insulin receptor substrate 53; insulin receptor substrate of 53 kDa; insulin receptor substrate p53; insulin receptor substrate p53/p58; Insulin receptor substrate protein of 53 kDa; insulin receptor tyrosine kinase 53 kDa substrate; Insulin receptor tyrosine kinase substrate protein p53; IRS-58; IRSp53; IRSp53/58; R75030 | |
Mouse | |
Affinity Chromatography | |
RUO | |
10458 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction