Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAIAP2L1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550993

Catalog No. PA550993


Add to cart

Description

Description

BAIAP2L1 Polyclonal Antibody detects endogenous levels of BAIAP2L1 protein.

May function as adapter protein. Involved in the formation of clusters of actin bundles. Plays a role in the reorganization of the actin cytoskeleton in response to bacterial infection. {ECO:0000269; PubMed:17430976, ECO:0000269; PubMed:19366662, ECO:0000269; PubMed:22921828}.
Specifications

Specifications

BAIAP2L1
Polyclonal
Unconjugated
BAIAP2L1
Liquid
BAIAP2L1
Synthesized peptide derived from the Internal region of human IRTKS.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
ELISA, Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Q9UHR4, Q9DBJ3, Q3KR97
IRTKS
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
304282, 55971, 66898
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit