Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Baicalein, Tocris Bioscience™
Click to view available options
Quantity:
50 mg
Description
Inhibitor of 5- and platelet 12-lipoxygenases (IC50 values are 9.5 and 0.12 μM respectively). Also inhibits Raf-mediated MEK-1 phosphorylation in C6 rat glioma cells and induces G1 and G2 cell cycle arrest by decreasing cdk1, cdk2, cyclin D2 and cyclin A expression. Inhibits production of inflammatory cytokines by inhibiting NF-κB activation.
Specifications
Quantity | 50 mg |
Formula Weight | 270.24 |
Percent Purity | >98% |
Chemical Name or Material | Baicalein |
Safety and Handling
Recommended Storage | Desiccate at -20°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction