Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BAK Rabbit anti-Human, Polyclonal, R&D Systems
Rabbit Polyclonal Antibody
Manufacturer: R&D Systems AF816SP
This item is not returnable.
View return policy
Description
BAK Polyclonal antibody AF816SP specifically detects BAK in Human samples. It is validated for Western BlottingSpecifications
BAK | |
LYOPH | |
Western Blot 1-4 ug/mL | |
Q16611 | |
Apoptosis regulator BAK, BAK1, BAK-LIKE, BAKMGC3887, BCL2-antagonist/killer 1, Bcl2-L-7, BCL2L7, BCL2L7bcl-2 homologous antagonist/killer, BCL2-like 7 protein, Bcl-2-like protein 7, CDN1, MGC117255, pro-apoptotic protein BAK | |
Rabbit | |
IgG | |
25 ug | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
Polyclonal | |
Detects human BAK in Western blots. This antibody does not detect mouse BAK. | |
Human |
Western Blot | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Affinity Purified | |
BAK1 | |
KLH-coupled human BAK synthetic peptide AAPADPEMVTLPLQPSSTMGC Accession # Q16611 | |
Immunogen affinity purified | |
RUO | |
Primary | |
578 | |
Reconstitute at 0.2 mg/mL in sterile PBS. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only