Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Baker-flex™, Aluminum Oxide IB-F, J.T. Baker™

Catalog No. p-5548222
Click to view available options
Quantity:
200 Sheets
25 Sheets
This item is not returnable. View return policy
This item is not returnable. View return policy

Flexible TLC Sheet

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Quantity 25 Sheets
Solubility Information Insoluble in water
IUPAC Name dialuminium(3+) trioxidandiide
Odor Odorless
Specific Gravity 1L = 4.00kg
Chemical Name or Material Aluminum Oxide IB-F
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.