Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Baker-flex™, Aluminum Oxide IB-F, J.T.Baker™

Click to view available options
Quantity:
25 sheets
200 sheets
1344-28-1
Al2O3
101.96
PNEYBMLMFCGWSK-UHFFFAOYSA-N
aluminium oxide,aluminum oxide,alpha-alumina,fasertonerde,abramant,abramax,abrarex,abrasit,aloxite,alundum
9989226
dialuminum;oxygen(2-)
[O-2].[O-2].[O-2].[Al+3].[Al+3]
1344-28-1
Al2O3
101.96
PNEYBMLMFCGWSK-UHFFFAOYSA-N
aluminium oxide,aluminum oxide,alpha-alumina,fasertonerde,abramant,abramax,abrarex,abrasit,aloxite,alundum
9989226
dialuminum;oxygen(2-)
[O-2].[O-2].[O-2].[Al+3].[Al+3]

Flexible TLC Sheet

Solubility Information Insoluble in water
Odor Odorless
Specific Gravity 1L = 4.00kg
Quantity 200 sheets
Chemical Name or Material Aluminum Oxide IB-F